Assay Details
Target Gene Details
Entrez Gene ID: | 12301 |
Gene Name: | calcyclin binding protein |
Gene Aliases: |
SIP |
Location: |
Chr.1:160202367-160212892 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacybp | NM_009786.2 | NP_033916.1 | ||
| AK013924.1 | ||||
| AK089043.1 | ||||
| AK145013.1 | ||||
| AK212050.1 | ||||
| AK215280.1 | ||||
| BC025948.1 | AAH25948.1 | |||
| U97327.1 | AAC16757.1 |
More Information
Set Membership: |
|

Genomic Map