Assay Details
Target Gene Details
Entrez Gene ID: | 12747 |
Gene Name: | CDC-like kinase 1 |
Gene Aliases: |
STY |
Location: |
Chr.1:58411988-58424088 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clk1 | NM_001042634.2 | NP_001036099.1 | ||
| NR_027853.1 | ||||
| NR_027854.1 | ||||
| AK031218.1 | BAE43276.1 | |||
| AK076199.1 | ||||
| AK135765.1 | BAE22647.1 | |||
| BC145905.1 | ||||
| BC145911.1 | ||||
| L29221.1 | AAA61485.1 | |||
| M38381.1 | AAA40151.1 | |||
| U11054.1 | AAA82185.1 | |||
| U21209.1 | AAC52257.1 | |||
| X57186.1 | CAA40473.1 |
More Information
Set Membership: |
|

Genomic Map