Assay Details
Target Gene Details
Entrez Gene ID: | 72978 |
Gene Name: | cornichon homolog 3 (Drosophila) |
Gene Aliases: |
2900075G08Rik, CNIH-3 |
Location: |
Chr.1:181352628-181460641 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnih3 | NM_001160211.1 | NP_001153683.1 | ||
| NM_001160212.1 | NP_001153684.1 | |||
| NM_028408.3 | NP_082684.1 | |||
| AK013789.1 | BAB28996.1 | |||
| AK033911.1 | BAC28511.1 | |||
| BC115640.1 | ||||
| BC115641.1 | ||||
| BG800984.1 |
More Information
Set Membership: |
|

Genomic Map