Assay Details
Target Gene Details
Entrez Gene ID: | 12825 |
Gene Name: | collagen, type III, alpha 1 |
Gene Aliases: |
AW550625, Col3a-1, MMS10-W, Ms10w, mKIAA4231 |
Location: |
Chr.1:45311538-45349706 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col3a1 | NM_009930.2 | NP_034060.2 | ||
| AK029212.1 | ||||
| AK041115.1 | BAC30826.1 | |||
| AK048546.1 | BAC33370.1 | |||
| AK147542.1 | BAE27985.1 | |||
| AK157762.1 | BAE34185.1 | |||
| AK159763.1 | ||||
| AK159920.1 | ||||
| AK159985.1 | ||||
| AK160048.1 | ||||
| AK160107.1 | BAE35633.1 | |||
| AK160818.1 | ||||
| AK220558.1 | BAD90325.1 | |||
| BC028248.1 | AAH28248.1 | |||
| BC043089.1 | AAH43089.1 | |||
| BC052398.1 | AAH52398.1 | |||
| BC058724.1 | AAH58724.1 | |||
| M18933.1 | AAA37338.1 |
More Information
Set Membership: |
|

Genomic Map