Assay Details
Target Gene Details
Entrez Gene ID: | 227292 |
Gene Name: | CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1 |
Gene Aliases: |
GIP, NLIIF, Nif3, SCP1 |
Location: |
Chr.1:74391609-74397285 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctdsp1 | NM_153088.2 | NP_694728.1 | ||
| AK089478.1 | ||||
| AK167390.1 | BAE39480.1 | |||
| AY028804.1 | AAK83555.1 | |||
| BC049184.1 | AAH49184.1 | |||
| BC065158.1 | AAH65158.1 | |||
| BC079638.1 | AAH79638.1 |

Genomic Map