Assay Details
Target Gene Details
Entrez Gene ID: | 77951 |
Gene Name: | cytochrome P450, family 20, subfamily a, polypeptide 1 |
Gene Aliases: |
A930011N14Rik, Cypm |
Location: |
Chr.1:60342800-60388060 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp20a1 | NM_030013.2 | NP_084289.1 | ||
| AK020848.1 | ||||
| AK053388.1 | BAC35373.1 | |||
| BC049147.1 | AAH49147.1 |
More Information
Set Membership: |
|

Genomic Map