Assay Details
Target Gene Details
Entrez Gene ID: | 104086 |
Gene Name: | cytochrome P450, family 27, subfamily a, polypeptide 1 |
Gene Aliases: |
1300013A03Rik, Cyp27 |
Location: |
Chr.1:74713146-74737896 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp27a1 | NM_024264.4 | NP_077226.2 | ||
| AK004977.1 | BAB23713.1 | |||
| BC002183.1 | AAH02183.1 | |||
| BC055028.1 | AAH55028.1 |
More Information
Set Membership: |
|

Genomic Map