Assay Details
Target Gene Details
Entrez Gene ID: | 26909 |
Gene Name: | exonuclease 1 |
Gene Aliases: |
5730442G03Rik, Msa |
Location: |
Chr.1:175880761-175911396 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exo1 | NM_012012.4 | NP_036142.2 | ||
| AK028728.1 | BAC26086.1 | |||
| AK166425.1 | BAE38768.1 | |||
| BC006671.1 | AAH06671.1 |
More Information
Set Membership: |
|

Genomic Map