Assay Details
Target Gene Details
Entrez Gene ID: | 213056 |
Gene Name: | family with sequence similarity 126, member B |
Gene Aliases: |
C130065N10Rik, D1Ertd53e, D630010C10 |
Location: |
Chr.1:58522806-58586333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam126b | NM_172513.3 | NP_766101.3 | ||
| AK052638.1 | BAC35073.1 | |||
| AK144602.1 | BAE25961.1 | |||
| AK168372.1 | BAE40304.1 | |||
| BC068302.1 | AAH68302.1 |

Genomic Map