Assay Details
Target Gene Details
Entrez Gene ID: | 620393 |
Gene Name: | family with sequence similarity 150, member A |
Gene Aliases: |
EG620393 |
Location: |
Chr.1:6359331-6394731 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam150a | NM_001195732.1 | NP_001182661.1 | ||
| AK087278.1 |
More Information
Set Membership: |
|

Genomic Map