Assay Details
Target Gene Details
Entrez Gene ID: | 381337 |
Gene Name: | family with sequence similarity 178, member B |
Gene Aliases: |
1700024G10Rik, Gm990 |
Location: |
Chr.1:36562696-36683190 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam178b | NM_001126046.1 | NP_001119518.1 | ||
| NM_201365.2 | NP_958753.2 | |||
| AK141223.1 | BAE24598.1 | |||
| BC050210.1 | AAH50210.1 |

Genomic Map