Assay Details
Target Gene Details
Entrez Gene ID: | 68226 |
Gene Name: | EF-hand calcium binding domain 2 |
Gene Aliases: |
1700073K01Rik, D830011E08Rik |
Location: |
Chr.1:178405881-178483250 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab2 | NM_026626.2 | NP_080902.1 | ||
| AK010880.1 | BAB27243.1 | |||
| AK015157.1 | BAB29727.1 | |||
| AK018894.1 | BAB31477.1 | |||
| AK019663.1 | BAB31827.1 | |||
| AK019802.1 | BAB31856.1 | |||
| BC025062.1 | AAH25062.1 |
More Information
Set Membership: |
|

Genomic Map