Assay Details
Target Gene Details
Entrez Gene ID: | 14103 |
Gene Name: | Fas ligand (TNF superfamily, member 6) |
Gene Aliases: |
APT1LG1, CD178, CD95-L, CD95L, Fas-L, Faslg, Tnfsf6, gld |
Location: |
Chr.1:161779664-161795538 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fasl | NM_001205243.1 | NP_001192172.1 | ||
| NM_010177.4 | NP_034307.1 | |||
| AF119335.1 | AAD52106.1 | |||
| AF288572.1 | AAG60016.1 | |||
| AK040139.1 | BAC30520.1 | |||
| AK141268.1 | BAE24625.1 | |||
| DQ846747.1 | ||||
| S76752.1 | AAB33780.1 | |||
| U06948.1 | AAA17800.1 | |||
| U10984.1 | AAA19778.1 | |||
| U58995.1 | AAB02915.1 |
More Information
Set Membership: |
|

Genomic Map