Assay Details
Target Gene Details
Entrez Gene ID: | 14660 |
Gene Name: | glutaminase |
Gene Aliases: |
6330442B14, AI314027, B230365M23Rik |
Location: |
Chr.1:52163449-52233232 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gls | NM_001081081.2 | NP_001074550.1 | ||
| NM_001113383.1 | NP_001106854.1 | |||
| AK032267.1 | ||||
| AK046297.1 | ||||
| AK048072.1 | ||||
| AK173039.1 | BAD32317.1 |
More Information
Set Membership: |
|

Genomic Map