Assay Details
Target Gene Details
Entrez Gene ID: | 16171 |
Gene Name: | interleukin 17A |
Gene Aliases: |
Ctla-8, Ctla8, IL-17, IL-17A, Il17 |
Location: |
Chr.1:20730905-20734496 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il17a | NM_010552.3 | NP_034682.1 | ||
| AK040420.1 | BAC30590.1 | |||
| BC119303.1 | ||||
| BC119309.1 |

Genomic Map