Assay Details
Target Gene Details
Entrez Gene ID: | 16177 |
Gene Name: | interleukin 1 receptor, type I |
Gene Aliases: |
CD121a, CD121b, IL-1R1, IL-iR, Il1r-1 |
Location: |
Chr.1:40225014-40316201 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1r1 | NM_001123382.1 | NP_001116854.1 | ||
| NM_008362.2 | NP_032388.1 | |||
| AK048550.1 | BAC33372.1 | |||
| AK054137.1 | BAC35666.1 | |||
| AK166301.1 | ||||
| BC109135.2 | AAI09136.1 | |||
| M20658.1 | AAA39279.1 |
More Information
Set Membership: |
|

Genomic Map