Assay Details
Target Gene Details
Entrez Gene ID: | 269180 |
Gene Name: | inositol polyphosphate-4-phosphatase, type I |
Gene Aliases: |
107kDa, 9630012D15, D130048C09Rik, R74740 |
Location: |
Chr.1:37299838-37410740 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp4a | NM_001290797.1 | NP_001277726.1 | ||
| NM_030266.4 | NP_084542.2 | |||
| AK149414.1 | BAE28860.1 | |||
| BY274715.1 |
More Information
Set Membership: |
|

Genomic Map