Assay Details
Target Gene Details
Entrez Gene ID: | 226432 |
Gene Name: | importin 9 |
Gene Aliases: |
0710008K06Rik, AI845704, C78347, Imp9, Ranbp9, mKIAA1192 |
Location: |
Chr.1:135382315-135430502 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ipo9 | NM_153774.1 | NP_722469.1 | ||
| AF273672.1 | AAK91127.1 | |||
| AF273673.1 | AAK91128.1 | |||
| AJ309238.1 | CAC69407.1 | |||
| AK003047.1 | ||||
| AK036687.1 | ||||
| AK173127.1 | BAD32405.1 | |||
| BC098508.1 | AAH98508.1 |
More Information
Set Membership: |
|

Genomic Map