Assay Details
Target Gene Details
Entrez Gene ID: | 240776 |
Gene Name: | potassium channel, subfamily T, member 2 |
Gene Aliases: |
E330038N15Rik, Slick |
Location: |
Chr.1:140245659-140613962 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnt2 | NM_001081027.2 | NP_001074496.2 |
More Information
Set Membership: |
|

Genomic Map