Assay Details
Target Gene Details
Entrez Gene ID: | 226519 |
Gene Name: | laminin, gamma 1 |
Gene Aliases: |
Lamb2 |
Location: |
Chr.1:153218922-153332786 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamc1 | NM_010683.2 | NP_034813.2 | ||
| AK141201.1 | BAE24583.1 | |||
| AK162773.1 | BAE37055.1 | |||
| J02930.1 | AAA39408.1 | |||
| J03484.1 | AAA39405.1 |
More Information
Set Membership: |
|

Genomic Map