Assay Details
Target Gene Details
Entrez Gene ID: | 226519 |
Gene Name: | laminin, gamma 1 |
Gene Aliases: |
Lamb2 |
Location: |
Chr.1:153218922-153332786 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamc1 | NM_010683.2 | 27 | 4766 | NP_034813.2 |
| AK141201.1 | 10 | 1285 | BAE24583.1 | |
| AK148199.1 | 7 | 873 | BAE28410.1 | |
| AK162773.1 | 8 | 1036 | BAE37055.1 | |
| BC056497.1 | 3 | 358 | AAH56497.1 | |
| J02930.1 | 27 | 4705 | AAA39408.1 | |
| J03484.1 | 27 | 4765 | AAA39405.1 | |
| X05211.1 | 3 | 350 | CAA28838.1 |
More Information
Set Membership: |
|

Genomic Map