Assay Details
Target Gene Details
Entrez Gene ID: | 13714 |
Gene Name: | ELK4, member of ETS oncogene family |
Gene Aliases: |
2310011G17Rik, A130026I01Rik, BB162516, SAP-1, Sap1 |
Location: |
Chr.1:132007234-132032612 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk4 | BC004798.1 | AAH04798.1 |
Target Gene Details
Entrez Gene ID: | 213006 |
Gene Name: | major facilitator superfamily domain containing 4 |
Gene Aliases: |
A230072B04, A930031D07Rik, AI850289 |
Location: |
Chr.1:132022806-132068062 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd4 | NM_172510.4 | NP_766098.2 | ||
| AK038882.1 | BAC30158.1 | |||
| AK044667.1 | 10 | 1615 | ||
| AK048692.1 | 9 | 1543 | BAC33423.1 | |
| AK162495.1 | BAE36946.1 | |||
| BC058790.1 | AAH58790.1 | |||
| FN434126.1 |
More Information
Set Membership: |
|

Genomic Map