Assay Details
Target Gene Details
Entrez Gene ID: | 107895 |
Gene Name: | mannoside acetylglucosaminyltransferase 5 |
Gene Aliases: |
4930471A21Rik, 5330407H02Rik, AI480971, GlcNAc-TV |
Location: |
Chr.1:127204977-127488334 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat5 | NM_145128.3 | NP_660110.2 | ||
| AK030402.1 | BAC26946.1 | |||
| AK036567.1 | ||||
| AK049587.1 | ||||
| BY752687.1 |
More Information
Set Membership: |
|

Genomic Map