Assay Details
Target Gene Details
Entrez Gene ID: | 227157 |
Gene Name: | membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4) |
Gene Aliases: |
Dlg6 |
Location: |
Chr.1:59120935-59163389 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp4 | NM_001164682.1 | 19 | 2091 | NP_001158154.1 |
| NM_145143.3 | 18 | 1454 | NP_660125.2 | |
| AB059357.1 | 17 | 1345 | BAB69494.1 | |
| AB059358.1 | 16 | 1327 | BAB69495.1 | |
| AJ748820.1 | 17 | 1335 | CAG38657.1 | |
| AK044682.1 | 19 | 2090 | BAC32031.1 | |
| BC061694.1 | 18 | 1371 | ||
| BC116723.1 | 13 | 974 |
More Information
Set Membership: |
|

Genomic Map