Assay Details
Target Gene Details
Entrez Gene ID: | 214854 |
Gene Name: | neuralized homolog 3 homolog (Drosophila) |
Gene Aliases: |
2010300P06Rik, Lincr |
Location: |
Chr.1:36264602-36274674 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neurl3 | NM_153408.2 | NP_700457.1 | ||
| AF321278.1 | AAN16205.1 | |||
| AK156207.1 | BAE33626.1 | |||
| BC056622.1 | AAH56622.1 |
More Information
Set Membership: |
|

Genomic Map