Assay Details
Target Gene Details
Entrez Gene ID: | 74229 |
Gene Name: | progestin and adipoQ receptor family member VIII |
Gene Aliases: |
1700019B16Rik, 3110001D06Rik |
Location: |
Chr.1:20890622-20939650 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paqr8 | NM_028829.3 | NP_083105.3 | ||
| AK029772.1 | BAC26609.1 | |||
| AK031969.1 | BAC27629.1 | |||
| AK048045.1 | BAC33221.1 | |||
| AK147376.1 | BAE27872.1 | |||
| AY424297.1 | AAR08385.1 | |||
| BC059813.1 | AAH59813.1 |
More Information
Set Membership: |
|

Genomic Map