Assay Details
Target Gene Details
Entrez Gene ID: | 19335 |
Gene Name: | RAB23, member RAS oncogene family |
Gene Aliases: |
AW545388, opb, opb2 |
Location: |
Chr.1:33719896-33742564 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab23 | NM_001159729.1 | NP_001153201.1 | ||
| NM_008999.4 | NP_033025.2 | |||
| AB232618.1 | BAF02880.1 | |||
| AK016493.1 | BAB30270.1 | |||
| AK019666.1 | ||||
| AK047068.1 | BAC32949.1 | |||
| AK082017.1 | ||||
| AK164057.1 | ||||
| BC025578.1 | AAH25578.1 | |||
| Z22821.1 | CAA80474.1 |

Genomic Map