Assay Details
Target Gene Details
Entrez Gene ID: | 226422 |
Gene Name: | RAB29, member RAS oncogene family |
Gene Aliases: |
Rab7l1 |
Location: |
Chr.1:131867210-131872887 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab29 | NM_144875.2 | NP_659124.2 | ||
| AB232625.1 | BAF02887.1 | |||
| AK079752.1 | ||||
| BC016133.1 | AAH16133.1 | |||
| BC029056.1 | AAH29056.1 | |||
| BG087764.2 |

Genomic Map