Assay Details
Target Gene Details
Entrez Gene ID: | 241113 |
Gene Name: | protein kinase, AMP-activated, gamma 3 non-catatlytic subunit |
Gene Aliases: |
AMPKg3L, AMPKg3S, Amkg3, Ampkg3 |
Location: |
Chr.1:74738922-74749590 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag3 | NM_153744.3 | NP_714966.1 | ||
| AF525500.1 | AAN47137.1 | |||
| AF525501.1 | AAN47138.1 | |||
| AK036585.1 | BAC29492.1 | |||
| BC116749.1 | ||||
| BC116777.1 |
More Information
Set Membership: |
|

Genomic Map