Assay Details
Target Gene Details
Entrez Gene ID: | 70247 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 1 |
Gene Aliases: |
2410026J11Rik, P112, S1 |
Location: |
Chr.1:86064619-86139295 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd1 | NM_027357.2 | NP_081633.1 | ||
| AK087947.1 | BAC40055.2 | |||
| AK153386.1 | BAE31950.1 | |||
| AK159123.1 | BAE34838.1 | |||
| AK159750.1 | ||||
| AK215835.1 | ||||
| BC022955.1 | ||||
| BC025802.1 | ||||
| BC031587.1 | AAH31587.1 | |||
| BC039937.1 | AAH39937.1 | |||
| BC089028.1 | AAH89028.1 | |||
| BC138526.1 | ||||
| BC138527.1 |
More Information
Set Membership: |
|

Genomic Map