Assay Details
Target Gene Details
Entrez Gene ID: | 19225 |
Gene Name: | prostaglandin-endoperoxide synthase 2 |
Gene Aliases: |
COX2, Cox-2, PGHS-2, PHS-2, Pghs2, TIS10 |
Location: |
Chr.1:150100124-150108017 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgs2 | NM_011198.3 | NP_035328.2 | ||
| AK049923.1 | BAC33986.1 | |||
| AK144726.1 | BAE26032.1 | |||
| AK144956.1 | BAE26154.1 | |||
| AK149820.1 | ||||
| AK166221.1 | BAE38639.1 | |||
| AK172161.1 | BAE42855.1 | |||
| BC052900.1 | AAH52900.1 | |||
| M64291.1 | AAA39924.1 | |||
| M88242.1 | AAA37740.1 | |||
| M94967.1 | AAA39918.1 |
More Information
Set Membership: |
|

Genomic Map