Assay Details
Target Gene Details
Entrez Gene ID: | 110895 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 4 |
Gene Aliases: |
AW990558, D730009J23Rik, NHE4 |
Location: |
Chr.1:40579715-40630731 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9a4 | NM_177084.3 | NP_796058.1 | ||
| AK085681.1 | BAC39504.1 | |||
| AK141534.1 | BAE24721.1 | |||
| AK170522.1 | BAE41856.1 | |||
| BC120543.1 |
More Information
Set Membership: |
|

Genomic Map