Assay Details
Target Gene Details
Entrez Gene ID: | 434484 |
Gene Name: | Sp140 nuclear body protein |
Gene Aliases: |
- |
Location: |
Chr.1:85600634-85645040 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp140 | NM_001013817.2 | NP_001013839.1 | ||
| BC069971.1 | AAH69971.1 |

Genomic Map