Assay Details
Target Gene Details
Entrez Gene ID: | 18173 |
Gene Name: | solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1 |
Gene Aliases: |
Bcg, Ity, Lsh, Nramp, Nramp1, ity |
Location: |
Chr.1:74375014-74386057 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc11a1 | NM_013612.2 | NP_038640.2 | ||
| AK090303.1 | ||||
| AK144237.1 | ||||
| AK171343.1 | 5 | 828 | ||
| AK171393.1 | BAE42430.1 | |||
| BC109137.1 | AAI09138.1 | |||
| BC109138.1 | AAI09139.1 | |||
| L13732.1 | AAA39838.1 | |||
| X75355.1 | CAA53102.1 |
More Information
Set Membership: |
|

Genomic Map