Assay Details
Target Gene Details
Entrez Gene ID: | 226781 |
Gene Name: | solute carrier family 30, member 10 |
Gene Aliases: |
E130106K10Rik, Gm212 |
Location: |
Chr.1:185454848-185468762 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc30a10 | NM_001033286.2 | NP_001028458.1 | ||
| AK053537.1 | ||||
| AK136932.1 | BAE23176.1 | |||
| BC108419.1 | AAI08420.1 |

Genomic Map