Assay Details
Target Gene Details
Entrez Gene ID: | 21956 |
Gene Name: | troponin T2, cardiac |
Gene Aliases: |
Tnt, cTnT |
Location: |
Chr.1:135836316-135852268 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnnt2 | NM_001130174.2 | NP_001123646.1 | ||
| NM_001130175.2 | NP_001123647.4 | |||
| NM_001130177.2 | NP_001123649.1 | |||
| NM_001130178.2 | NP_001123650.1 | |||
| NM_001130179.2 | NP_001123651.4 | |||
| NM_001130180.2 | NP_001123652.1 | |||
| NM_001130181.2 | NP_001123653.4 | |||
| NM_011619.3 | NP_035749.1 | |||
| AK086260.1 | 1 | 650 | ||
| AK146945.1 | BAE27554.1 | |||
| AK163786.1 | BAE37494.1 | |||
| AK164592.1 | ||||
| AK168393.1 | BAE40318.1 | |||
| AK169061.1 | ||||
| BC063753.1 | AAH63753.1 | |||
| BQ554639.1 | ||||
| BY060272.1 | ||||
| BY064679.1 | ||||
| BY067092.1 | ||||
| L47549.1 | AAA85345.1 | |||
| L47550.1 | AAA85346.1 | |||
| L47551.1 | AAA85347.1 | |||
| L47552.1 | AAA85348.1 | |||
| L47599.1 | AAA85351.1 | |||
| L47600.1 | AAA85352.1 |
More Information
Set Membership: |
|

Genomic Map