Assay Details
Target Gene Details
Entrez Gene ID: | 27878 |
Gene Name: | transcriptional adaptor 1 |
Gene Aliases: |
2900026B15Rik, D1Ertd251e, Staf42, Tada1l |
Location: |
Chr.1:166379167-166393621 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tada1 | NM_030245.3 | NP_084521.1 | ||
| AK013599.1 | ||||
| AK050649.1 | ||||
| AK151132.1 | BAE30140.1 | |||
| BC002307.1 | AAH02307.1 | |||
| BC027337.1 | AAH27337.1 |

Genomic Map