Assay Details
Target Gene Details
Entrez Gene ID: | 21346 |
Gene Name: | transgelin 2 |
Gene Aliases: |
2700094C18Rik, SM22beta, Sm22B, Sm22a |
Location: |
Chr.1:172500246-172507375 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tagln2 | NM_178598.2 | NP_848713.1 | ||
| AK151576.1 | BAE30516.1 | |||
| AK167395.1 | BAE39484.1 | |||
| BC049861.1 | 1 | 163 |
More Information
Set Membership: |
|

Genomic Map