Assay Details
Target Gene Details
Entrez Gene ID: | 170706 |
Gene Name: | transmembrane protein 37 |
Gene Aliases: |
AI173373, AW540203, Pr, Pr1 |
Location: |
Chr.1:120067377-120074069 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem37 | NM_019432.2 | NP_062305.1 | ||
| AK002424.1 | BAB22091.1 | |||
| AK050134.1 | BAC34086.1 | |||
| AK086093.1 | BAC39608.1 | |||
| BC024613.1 | AAH24613.1 |
More Information
Set Membership: |
|

Genomic Map