Assay Details
Target Gene Details
Entrez Gene ID: | 29819 |
Gene Name: | staufen (RNA binding protein) homolog 2 (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.1:16228803-16520181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stau2 | NM_001111272.1 | NP_001104742.1 | ||
| NM_025303.3 | NP_079579.2 | |||
| AF459099.1 | AAN37928.1 | |||
| AF459100.1 | AAN37929.1 | |||
| AK015087.1 | BAB29708.1 | |||
| AK031102.1 | ||||
| AK171076.1 | ||||
| BC010300.1 | AAH10300.1 | |||
| BC025118.1 | AAH25118.1 |
More Information
Set Membership: |
|

Genomic Map