Assay Details
Target Gene Details
Entrez Gene ID: | 68428 |
Gene Name: | STEAP family member 3 |
Gene Aliases: |
1010001D01Rik, Tsap6, pHyde |
Location: |
Chr.1:120226414-120271082 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Steap3 | NM_001085409.1 | NP_001078878.1 | ||
| NM_133186.3 | NP_573449.2 | |||
| AK028961.1 | ||||
| AK078769.1 | BAC37383.1 | |||
| AK167028.1 | BAE39201.1 | |||
| AK171237.1 | BAE42333.1 | |||
| AY029586.1 | AAK50539.1 | |||
| BB857879.1 | ||||
| BC037435.1 | AAH37435.1 | |||
| BY718114.1 |

Genomic Map