Assay Details
Target Gene Details
Entrez Gene ID: | 67534 |
Gene Name: | tubulin tyrosine ligase-like family, member 4 |
Gene Aliases: |
4632407P03Rik, AI451681, mKIAA0173 |
Location: |
Chr.1:74661645-74701832 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll4 | NM_001014974.1 | NP_001014974.1 | ||
| AK129075.1 | BAC97885.1 | |||
| AM690747.1 | ||||
| BC044790.1 | AAH44790.1 | |||
| BC085151.1 | AAH85151.1 |
More Information
Set Membership: |
|

Genomic Map