Assay Details
Target Gene Details
Entrez Gene ID: | 68002 |
Gene Name: | succinate dehydrogenase complex assembly factor 4 |
Gene Aliases: |
1110058L19Rik, 1700001E18Rik, C6orf57 |
Location: |
Chr.1:23995939-24005640 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdhaf4 | NM_026503.3 | NP_080779.2 | ||
| AK003789.1 | BAC25055.1 | |||
| AK004301.1 | BAB23255.1 | |||
| AK007909.1 | BAB25342.1 | |||
| AK015824.1 | BAB29991.1 | |||
| BC028981.1 | AAH28981.1 |
More Information
Set Membership: |
|

Genomic Map