Assay Details
Target Gene Details
Entrez Gene ID: | 78258 |
Gene Name: | maestro heat-like repeat family member 9 |
Gene Aliases: |
4921528O07Rik, Armc11 |
Location: |
Chr.1:163024302-163085670 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mroh9 | NM_030071.1 | NP_084347.1 | ||
| AK019544.1 | BAB31789.1 |

Genomic Map