Assay Details
Target Gene Details
Entrez Gene ID: | 74895 |
Gene Name: | coiled-coil domain containing 181 |
Gene Aliases: |
4930455F23Rik, AI427889 |
Location: |
Chr.1:164275598-164287847 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc181 | NM_029115.3 | NP_083391.2 | ||
| AK076640.1 | BAC36430.1 | |||
| BC048086.1 | AAH48086.1 |

Genomic Map