Assay Details
Target Gene Details
Entrez Gene ID: | 74205 |
Gene Name: | acyl-CoA synthetase long-chain family member 3 |
Gene Aliases: |
2610510B12Rik, Acs3, C85929, Facl3, Pro2194 |
Location: |
Chr.1:78657825-78707743 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsl3 | NM_001033606.2 | 3 | 747 | NP_001028778.2 |
| NM_001136222.1 | 2 | 329 | NP_001129694.1 | |
| NM_028817.3 | 4 | 887 | NP_083093.2 | |
| AK012088.1 | 3 | 690 | BAB28022.1 | |
| AK160839.1 | 2 | 273 | ||
| AK161708.1 | 4 | 888 | BAE36543.1 | |
| AK166723.1 | 4 | 825 | ||
| AK195074.1 | 2 | 107 | ||
| BC031529.1 | 2 | 472 | AAH31529.1 |
More Information
Set Membership: |
|

Genomic Map