Assay Details
Target Gene Details
Entrez Gene ID: | 619289 |
Gene Name: | regulatory factor X 8 |
Gene Aliases: |
4933400N17Rik |
Location: |
Chr.1:39665299-39720999 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx8 | NM_001145660.1 | NP_001139132.1 | ||
| AK133372.1 |
More Information
Set Membership: |
|

Genomic Map