Assay Details
Target Gene Details
Entrez Gene ID: | 74117 |
Gene Name: | ARP3 actin-related protein 3 |
Gene Aliases: |
1200003A09Rik, Arp3 |
Location: |
Chr.1:125392905-125435727 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr3 | NM_001205385.1 | NP_001192314.1 | ||
| NM_001205386.1 | NP_001192315.1 | |||
| NM_023735.2 | NP_076224.1 | |||
| AK004554.1 | BAB23368.1 | |||
| AK083343.1 | BAC38876.1 | |||
| AK145536.1 | BAE26491.1 | |||
| AK150487.1 | BAE29603.1 | |||
| AK150589.1 | BAE29683.1 | |||
| AK150991.1 | ||||
| AK152865.1 | ||||
| AK153079.1 | BAE31704.1 | |||
| AK153150.1 | BAE31760.1 | |||
| AK153204.1 | BAE31804.1 | |||
| AK159135.1 | BAE34847.1 | |||
| AK159668.1 | BAE35272.1 | |||
| AK159775.1 | BAE35360.1 | |||
| AK160047.1 | BAE35589.1 | |||
| AK166529.1 | BAE38833.1 | |||
| AK166586.1 | BAE38875.1 | |||
| AK167148.1 | BAE39291.1 | |||
| AK167891.1 | BAE39903.1 | |||
| AK168568.1 | BAE40438.1 | |||
| AK168771.1 | BAE40607.1 | |||
| AK187649.1 | ||||
| AK191050.1 | ||||
| AK211935.1 | ||||
| BC005557.1 | AAH05557.1 | |||
| BC080806.1 | AAH80806.1 | |||
| BC108386.1 | AAI08387.1 |
More Information
Set Membership: |
|

Genomic Map