Assay Details
Target Gene Details
Entrez Gene ID: | 23792 |
Gene Name: | a disintegrin and metallopeptidase domain 23 |
Gene Aliases: |
AW046396, MDC3 |
Location: |
Chr.1:63445885-63596515 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam23 | NM_001177600.1 | 22 | 2315 | NP_001171071.1 |
| NM_011780.2 | 22 | 2315 | NP_035910.1 | |
| AB009673.1 | 22 | 2087 | BAA83381.1 | |
| AK029301.1 | 20 | 1630 | ||
| AK034022.1 | 22 | 2310 | BAC28550.1 | |
| AK046677.1 | 22 | 2329 | ||
| AY545640.1 | AAS49900.1 | |||
| AY545641.1 | AAS49901.1 |
More Information
Set Membership: |
|

Genomic Map