Assay Details
Target Gene Details
Entrez Gene ID: | 11566 |
Gene Name: | adenylosuccinate synthetase, non muscle |
Gene Aliases: |
AI314886, AS, Adss2 |
Location: |
Chr.1:177763178-177796694 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adss | NM_007422.3 | 6 | 639 | NP_031448.2 |
| AK004877.1 | 6 | 592 | BAB23635.1 | |
| AK010263.1 | 6 | 614 | BAB26805.1 | |
| AK028060.1 | 6 | 639 | BAC25730.1 | |
| AK043273.1 | 6 | 632 | BAC31512.1 | |
| AK148420.1 | 6 | 621 | BAE28543.1 | |
| AK170279.1 | 6 | 625 | BAE41682.1 | |
| BC139404.1 | 6 | 551 | ||
| BC139417.1 | 6 | 551 | ||
| L24554.1 | 6 | 606 | AAA19727.1 |
More Information
Set Membership: |
|

Genomic Map